Scissors, Vannas, Fine, Mortech
Scissors, Vannas, Fine, Mortech
Supplier:   Mopec
Scissors, Vannas, Fine, Mortech
Supplier:  Mopec
Certificates
Scissors, Vannas, Fine, Mortech
Scissors, Vannas, Fine, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Designed to maximize visibility and have a long taper ideal for precise cuts.
  • All micro scissors are made from stainless steel.
Available in multiple lengths.
Product Family Options
Product Information
  • Longueur
    Availability
    Price
  • 3.25"
    Log in for availability
    Log in for availability
    Each
    Each - $152.03
    $152.03
    Each
       $152.03
    Supplier Number: AA437
  • 4"
    Log in for availability
    Log in for availability
    Each
    Each - $164.50
    $164.50
    Each
       $164.50
    Supplier Number: AA438