Scissors, Tungsten Carbide, Iris, Mortech | Avantor
Scissors, Tungsten Carbide, Iris, Mortech
Supplier:   Mopec
Scissors, Tungsten Carbide, Iris, Mortech
Supplier:  Mopec
Certificates
Scissors, Tungsten Carbide, Iris, Mortech
Scissors, Tungsten Carbide, Iris, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The unique tungsten carbide cutting edge provides effortless, yet accurate cuts through the toughest of tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex post-mortem dissection cases.
  • Premium grade stainless steel
  • Sharp/sharp points
Available with straight or curved edges.
Product Family Options
Informations sur le produit
  • Longueur
    Curvature
    Availability
    Price
  • 4.5"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 596,06 $
    596,06 $
    Each
       596,06 $
    Supplier Number: AA405
  • 4.5"
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 616,03 $
    616,03 $
    Each
       616,03 $
    Supplier Number: AA406