Scissors, Tungsten Carbide, Mortech
Scissors, Tungsten Carbide, Mortech
Supplier:   Mopec
Scissors, Tungsten Carbide, Mortech
Supplier:  Mopec
Certificates
Scissors, Tungsten Carbide, Mortech
Scissors, Tungsten Carbide, Mortech
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The unique tungsten carbide cutting edge provides effortless, yet accurate cuts through the toughest of tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex post-mortem dissection cases.
  • Tungsten Carbide
Available with straight or curved edges, and either sharp/sharp or sharp/blunt tips.
Product Family Options
Product Information
  • Longueur
    Blade Type
    Curvature
    Availability
    Price
  • 5.5"
    Sharp/Blunt
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $581.08
    $581.08
    Each
       $581.08
    Supplier Number: AA401
  • 5.5"
    Sharp/Sharp
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $581.08
    $581.08
    Each
       $581.08
    Supplier Number: AA402
  • 5.5"
    Sharp/Sharp
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $892.95
    $892.95
    Each
       $892.95
    Supplier Number: AA403
  • 5.5"
    Sharp/Blunt
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $596.06
    $596.06
    Each
       $596.06
    Supplier Number: AA404