Dissecting Kit, Student, Mortech
Dissecting Kit, Student, Mortech
Catalog # 76548-518
Supplier:   Mopec
Supplier #:  AA330
Dissecting Kit, Student, Mortech
Catalog # 76548-518
Supplier:  Mopec
Dissecting Kit, Student, Mortech
Dissecting Kit, Student, Mortech
Price
$339.10
Log in for availability
Each
Product Details & Documents
Kit includes 13 standard instruments in elasticized straps to suit a variety of post-mortem dissection needs.
 : Vinyl case kit includes 13 instruments to suit a variety of post-mortem dissection needs: 1- #4 knife handle; 1- Mayo Hegar needle holder, 6", 1- dressing forceps, serrated, 5.5", 1- tissue forceps, 1×2 teeth, 5.5", 1- grooved Dir., tongue tie, probe tip 5.5", 1- probe with eye, 5.5", 1- Kelly hemostatic forceps, Str., 5.5", 1- Kelly hemostatic forceps, Cvd., 5.5", 1- O.R. scissors, Str., Sh/Bl, 5.5", 1- O.R. scissors, Cvd., Sh/Bl, 5.5", 1- Mayo dissecting scissors, Str., 5.5", 1- Mayo dissecting scissors, Cvd., 5.5", 1- Halstead mosquito forceps, Str., 5".