ContactUsLinkComponent
Dissecting Kit, Anatomy, Mortech
Catalog # 76548-510
undefined
Dissecting Kit, Anatomy, Mortech
Catalog # 76548-510
Supplier Number:  AA305
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. no.:
    76548-510
Specifications
About this item
Includes 10 standard instruments in a vinyl case with elasticized straps to perform a variety of post-mortem dissection needs.
Delivery: Kit contains one of each instrument listed: model AA008 dissecting scissors, straight, 5.5"; BA021 Mayo scissors, curved, 5.5", AB003 dressing forceps, 5.5", AB011 tissue forceps, 1×2 Teeth, 5.5", AC003 Kelley forceps, curved, 5.5", AD004 grooved director, plain end, 5.5", AD029 triple chain hook, AD041 Mayo-hegar needle holder, 6", AJ101 knife handle #4.