Scissors, Delicate, Double Curved, Sharp, Mortech | Avantor
Scissors, Delicate, Double Curved, Sharp, Mortech
4.625"
Catalog # 76548-496
Supplier:   Mopec
Fournisseur #:  AA154
Scissors, Delicate, Double Curved, Sharp, Mortech
Catalog # 76548-496
Supplier:  Mopec
Scissors, Delicate, Double Curved, Sharp, Mortech
Scissors, Delicate, Double Curved, Sharp, Mortech
4.625"
Price
$364.08
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Finger-friendly ribbon handle. Extra sharp with single serrated edge. Double curvature of these scissors makes post-mortem dissection easier in hard to reach areas or where a clear view is not always available.
  • Premium grade stainless steel
  • Curved cutting edge
  • Sharp, one side of scissor edge is serrated
Spécifications
  • Longueur:
    4.625"