Scissors, Superfine Micro, Mortech | Avantor
Scissors, Superfine Micro, Mortech
Supplier:   Mopec
Scissors, Superfine Micro, Mortech
Supplier:  Mopec
Certificates
Scissors, Superfine Micro, Mortech
Scissors, Superfine Micro, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Delicate scissors with a fine point for small, very precise, post-mortem dissection.
  • Premium grade stainless steel
  • Superfine sharp tips
Available with straight or curved edges.
Product Family Options
Informations sur le produit
  • Longueur
    Curvature
    Availability
    Price
  • 3.25"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,184.79
    $1,184.79
    Each
       $1,184.79
    Supplier Number: AA152
  • 3.25"
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,202.26
    $1,202.26
    Each
       $1,202.26
    Supplier Number: AA153