Scissors, Micro Iris, Mortech | Avantor
Scissors, Micro Iris, Mortech
Supplier:   Mopec
Scissors, Micro Iris, Mortech
Supplier:  Mopec
Certificates
Scissors, Micro Iris, Mortech
Scissors, Micro Iris, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Delicate scissors excellent for small, very precise, post-mortem dissection.
  • Premium grade stainless steel
  • Sharp points
Available with straight or curved edges.
Product Family Options
Informations sur le produit
  • Longueur
    Curvature
    Availability
    Price
  • 4"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,015.17
    $1,015.17
    Each
       $1,015.17
    Supplier Number: AA150
  • 4"
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,184.79
    $1,184.79
    Each
       $1,184.79
    Supplier Number: AA151