Scissors, Iris Dissection, Mortech
Scissors, Iris Dissection, Mortech
Supplier:   Mopec
Scissors, Iris Dissection, Mortech
Supplier:  Mopec
Certificates
Scissors, Iris Dissection, Mortech
Scissors, Iris Dissection, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Also referred to as the 'Ribbon Scissor' because of the unique and finger-friendly, comfort handle. With extra wide finger rings offers comfortable post-mortem dissection. This precise and delicate scissor is made of the highest quality German stainless steel. Extra sharp tips allow for intricate work.
  • High grade stainless steel
  • Extra sharp tips
Available with straight or curved tips.
Product Family Options
Product Information
  • Longueur
    Curvature
    Availability
    Price
  • 4"
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $361.59
    $361.59
    Each
       $361.59
    Supplier Number: AA146
  • 4"
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $418.92
    $418.92
    Each
       $418.92
    Supplier Number: AA147