Scissors, Doyen Abdominal, Mortech | Avantor
Scissors, Doyen Abdominal, Mortech
Supplier:   Mopec
Scissors, Doyen Abdominal, Mortech
Supplier:  Mopec
Certificates
Scissors, Doyen Abdominal, Mortech
Scissors, Doyen Abdominal, Mortech
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
General purpose, blunt end post-mortem dissection scissors with extended shaft length. Curved option is available.
  • Stainless steel
  • Blunt points
Available in standard or premium grade.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Curvature
    Availability
    Price
  • Standard Grade
    7"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $134.53
    $134.53
    Each
       $134.53
    Supplier Number: AA035
  • Standard Grade
    7"
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $134.53
    $134.53
    Each
       $134.53
    Supplier Number: AA036
  • Premium Grade
    7"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $339.10
    $339.10
    Each
       $339.10
    Supplier Number: AA135
  • Premium Grade
    7"
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $351.60
    $351.60
    Each
       $351.60
    Supplier Number: AA136