Scissors, Enterotomy, Mortech | Avantor
Scissors, Enterotomy, Mortech
Supplier:   Mopec
Scissors, Enterotomy, Mortech
Supplier:  Mopec
Certificates
Scissors, Enterotomy, Mortech
Scissors, Enterotomy, Mortech
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Excellent scissors for small cases.
  • Standard grade stainless steel
  • Angled end cutting edge
  • Lower probe point
Available in right handed and left handed options.
Product Family Options
Informations sur le produit
  • Longueur
    Handle
    Availability
    Price
  • 3.5"
    Right-handed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $42.29
    $42.29
    Each
       $42.29
    Supplier Number: AA031
  • 4.5"
    Right-handed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $52.25
    $52.25
    Each
       $52.25
    Supplier Number: AA032
  • 5.5"
    Right-handed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $57.22
    $57.22
    Each
       $57.22
    Supplier Number: AA033
  • 5.5"
    Left-handed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $89.66
    $89.66
    Each
       $89.66
    Supplier Number: AA043