Scissors, Iris, Angled, Mortech | Avantor
Scissors, Iris, Angled, Mortech
Supplier:   Mopec
Scissors, Iris, Angled, Mortech
Supplier:  Mopec
Certificates
Scissors, Iris, Angled, Mortech
Scissors, Iris, Angled, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for small, precise, post-mortem dissection.
  • Stainless steel
  • Angled end cutting edge
  • Sharp points
Available in standard or premium grade.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Standard Grade
    4.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $69.65
    $69.65
    Each
       $69.65
    Supplier Number: AA030
  • Premium Grade
    4.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $246.84
    $246.84
    Each
       $246.84
    Supplier Number: AA130