Scissors, Metzenbaum, Curved, Mortech
À propos de cet article
These scissors offer a unique combination of slim, blunt, curved tips and thin, delicate shafts for post-mortem dissection with minimal damage to surrounding tissues. Variety of lengths and grades.
- Stainless steel
- Curved cutting edge
- Narrow blunt points
Available in standard or premium grade.
8 Options Available Below
Product Details & Documents
These scissors offer a unique combination of slim, blunt, curved tips and thin, delicate shafts for post-mortem dissection with minimal damage to surrounding tissues. Variety of lengths and grades.
- Stainless steel
- Curved cutting edge
- Narrow blunt points
Available in standard or premium grade.
Product Family Options
Informations sur le produit
- DésignationLongueurAvailabilityPrice
- Supplier Number: AA025
- Supplier Number: AA026
- Supplier Number: AA039
- Supplier Number: AA040
- Supplier Number: AA125
- Supplier Number: AA126
- Supplier Number: AA139
- Supplier Number: AA140