Scissors, Metzenbaum, Straight, Mortech
À propos de cet article
These scissors offer a unique combination of slim, blunt tips and thin, delicate shafts for post-mortem dissection with minimal damage to surrounding tissues. Variety of lengths and grades.
- Stainless steel
- Straight cutting edge
- Narrow blunt points
Available in standard or premium grade.
8 Options Available Below
Product Details & Documents
These scissors offer a unique combination of slim, blunt tips and thin, delicate shafts for post-mortem dissection with minimal damage to surrounding tissues. Variety of lengths and grades.
- Stainless steel
- Straight cutting edge
- Narrow blunt points
Available in standard or premium grade.
Product Family Options
Informations sur le produit
- DésignationLongueurAvailabilityPrice
- Supplier Number: AA023
- Supplier Number: AA024
- Supplier Number: AA037
- Supplier Number: AA038
- Supplier Number: AA123
- Supplier Number: AA124
- Supplier Number: AA137
- Supplier Number: AA138