Scissors, Mayo Dissection, Curved, Mortech | Avantor
Scissors, Mayo Dissection, Curved, Mortech
Supplier:   Mopec
Scissors, Mayo Dissection, Curved, Mortech
Supplier:  Mopec
Certificates
Scissors, Mayo Dissection, Curved, Mortech
Scissors, Mayo Dissection, Curved, Mortech
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Slightly blunt tips assure precise, controlled and safe post-mortem dissection with minimal damage to surrounding tissue.
  • Stainless steel
  • Curved cutting edge
  • Blunt tips
Available in standard or premium grade.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Standard Grade
    5.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $67.22
    $67.22
    Each
       $67.22
    Supplier Number: AA021
  • Standard Grade
    6.75"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $99.65
    $99.65
    Each
       $99.65
    Supplier Number: AA022
  • Standard Grade
    9"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $119.59
    $119.59
    Each
       $119.59
    Supplier Number: AA050
  • Premium Grade
    5.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $179.47
    $179.47
    Each
       $179.47
    Supplier Number: AA121
  • Premium Grade
    6.75"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $216.90
    $216.90
    Each
       $216.90
    Supplier Number: AA122