Scissors, Mayo Dissection, Straight, Mortech
À propos de cet article
Slightly blunt tips assure controlled, precise and safe post-mortem dissection with minimal damage to surrounding tissue.
- Stainless steel
- Straight cutting edge
- Blunt tips
Available in standard or premium grade.
6 Options Available Below
Product Details & Documents
Slightly blunt tips assure controlled, precise and safe post-mortem dissection with minimal damage to surrounding tissue.
- Stainless steel
- Straight cutting edge
- Blunt tips
Available in standard or premium grade.
Product Family Options
Informations sur le produit
- DésignationLongueurAvailabilityPrice
- Supplier Number: AA019
- Supplier Number: AA020
- Supplier Number: AA049
- Supplier Number: AA119
- Supplier Number: AA120
- Supplier Number: AA221