Scissors, Dissection, Blunt/Blunt, Straight, Mortech
 :   Mopec
Scissors, Dissection, Blunt/Blunt, Straight, Mortech
 :  Mopec
Scissors, Dissection, Blunt/Blunt, Straight, Mortech
Scissors, Dissection, Blunt/Blunt, Straight, Mortech
These blunt tipped scissors allow for safe post-mortem dissection procedures.
  • Stainless steel
  • Straight cutting edge
  • Blunt/blunt tips
Available in standard or premium grade.
Product Family Options
  • Désignation
    Longueur
  • Standard Grade
    4.5"
    Each
    Each - $42.29
    $42.29
    Each
       $42.29
     : AA013
  • Standard Grade
    5.5"
    Each
    Each - $52.25
    $52.25
    Each
       $52.25
     : AA014
  • Standard Grade
    6.5"
    Each
    Each - $57.22
    $57.22
    Each
       $57.22
     : AA015
  • Premium Grade
    4.5"
    Each
    Each - $194.46
    $194.46
    Each
       $194.46
     : AA113
  • Premium Grade
    5.5"
    Each
    Each - $199.45
    $199.45
    Each
       $199.45
     : AA114
  • Premium Grade
    6.5"
    Each
    Each - $231.85
    $231.85
    Each
       $231.85
     : AA115