Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Supplier:   Mopec
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Supplier:  Mopec
Certificates
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These scissors are designed with one sharp tip, one blunt tip and a curved cutting edge. Ideal for a variety of post-mortem dissection needs.
  • Stainless steel
  • Curved cutting edge
  • Sharp/blunt tips
Available in standard or premium grade.
Product Family Options
Product Information
  • Désignation
    Longueur
    Availability
    Price
  • Standard Grade
    4.5"
    Log in for availability
    Log in for availability
    Each
    Each - $42.29
    $42.29
    Each
       $42.29
    Supplier Number: AA010
  • Standard Grade
    5.5"
    Log in for availability
    Log in for availability
    Each
    Each - $52.25
    $52.25
    Each
       $52.25
    Supplier Number: AA011
  • Standard Grade
    6.5"
    Log in for availability
    Log in for availability
    Each
    Each - $57.22
    $57.22
    Each
       $57.22
    Supplier Number: AA012
  • Premium Grade
    4.5"
    Log in for availability
    Log in for availability
    Each
    Each - $209.45
    $209.45
    Each
       $209.45
    Supplier Number: AA110
  • Premium Grade
    5.5"
    Log in for availability
    Log in for availability
    Each
    Each - $229.35
    $229.35
    Each
       $229.35
    Supplier Number: AA111
  • Premium Grade
    6.5"
    Log in for availability
    Log in for availability
    Each
    Each - $256.84
    $256.84
    Each
       $256.84
    Supplier Number: AA112