Scissors, Dissection, Sharp/Blunt, Straight, Mortech
À propos de cet article
These scissors are designed with one sharp tip, one blunt tip and a straight cutting edge. Ideal for a variety of post-mortem dissection needs.
- Stainless steel
- Straight cutting edge
- Sharp/blunt tips
Available in standard or premium grade.
6 Options Available Below
Product Details & Documents
These scissors are designed with one sharp tip, one blunt tip and a straight cutting edge. Ideal for a variety of post-mortem dissection needs.
- Stainless steel
- Straight cutting edge
- Sharp/blunt tips
Available in standard or premium grade.
Product Family Options
Informations sur le produit
- DésignationLongueurAvailabilityPrice
- Supplier Number: AA007
- Supplier Number: AA008
- Supplier Number: AA009
- Supplier Number: AA107
- Supplier Number: AA108
- Supplier Number: AA109