Dissecting Kit
Dissecting kit
470339-188
: UNDSET08-W
Stainless steel dissecting set with hard protective case.
- Hard protective case
- Stainless steel instruments
A set of 8 dissection instruments in a hard plastic case.
Kit includes: Dissecting Scissors (1); Dissecting Forceps (1); Scalpel with Replaceable Blade, No. 22 (1); Teasing Needle, Plastic Handle, straight (1); Teasing Needle, Plastic Handle, curved (2); Dropping Pipet (1); Transparent Vinyl Ruler, 6" (1); T-Pins (6).
- Désignation:Dissecting kit