Equal Volume Density "Squares"
Equal volume metal squares, set of 4
470339-454
: SGVS04
Square column metal set for density and specific gravity studies.
- Set of 4 equal volume rectangular prisms
- Includes: Aluminum, Copper, Steel and Brass
- Dimensions: 1.26×1.26×5 cm (0.5×0.5×2")
- Storage box and activity guide included
- Ideal for lessons around density and specific gravity
Square specimen set is used for density and specific gravity studies. The square shape makes measurement and volume calculations easy for students. Set of four includes aluminum, copper, steel, and brass cylinders. Includes a storage box and activity guide.
- Désignation:Equal volume metal squares, set of 4