United Quiet Air Source with Variable Output
Catalogue # 470339-012
Fournisseur: United Scientific Supplies
Spécifications
- Description:Quiet air source with variable output
- Cat. no.:470339-012
Spécifications
À propos de cet article
Low noise air source for air tracks.
Low noise (<58 dB) air source - quieter than normal conversation
- Ideal accessory for air tracks, air tables, and other pneumatic equipment in physics labs
This low noise (<58 dB) air source is an ideal accessory for air tracks, air tables, and other pneumatic equipment in physics labs. The 250 W motor produces clean air output with a flow rate of 35 m³/hour at a 0.70 kPa overpressure.