Human Skull Model
Human skull model
Catalog # 470339-942
Supplier: United Scientific Supplies
Fournisseur #: PSM001
Jump to:
Product Details & Documents
Life sized human skull with three removable teeth.
- Life-sized human skull model, ideal for introductory anatomical studies
- Detailed key included
This life-sized skull model is excellent for introductory anatomy courses. The model shows excellent detail with a removable molar, incisor, and canine tooth for further study of the tooth roots. The mandible is spring mounted and the sectioned calvarium is detachable.
Spécifications
- Désignation:Human skull model