Hand Microtome
Hand microtome
Catalog # 470339-070
Supplier: United Scientific Supplies
Supplier #: MCT001
Jump to:
Product Details & Documents
Easy to use microtome to prepare slide sections.
- Tempered steel razor cuts sections to 10 μm (0.01 mm) with a few simple twists
- Clamping mechanism secures objects of various shapes and sizes
- Includes wooden storage box, with two glass overlays for stage
- Safe for classroom use; instructions included
Safe for use in introductory biology and histology classrooms, this hand-operated microtome allows students to precisely cut sample sections as small as 10 microns (0.01 mm) with a few simple twists. The unit ships in a durable wooden storage box, with two glass overlays to serve as a stage.
Specifications
- Désignation:Hand microtome