Deluxe Micrometer
Deluxe micrometer
Catalog # 470339-068
Supplier: United Scientific Supplies
: MCRDLX
Jump to:
Product Details & Documents
Precision micrometer with a resolution of 0.01 mm.
- Made of hardened steel for precision and durability
- Features ratchet thimble and a locking lever
- Instructions included
This micrometer is made of hardened steel for precision and durability. Its range is 0 to 25 mm, and it has a precision rotary scale with a resolution of 0.01 mm, a ratchet thimble, and a locking lever.
Spécifications
- Désignation:Deluxe micrometer