Human Skeleton Model, Life-Sized
Human Skeleton Model, Life-Sized
Human skeleton model, life-sized
Catalog # 470339-164
Supplier #:  HSLS01
Human Skeleton Model, Life-Sized
Catalog # 470339-164
Human Skeleton Model, Life-Sized
Human Skeleton Model, Life-Sized
Human skeleton model, life-sized
Price
$599.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Life sized human skeleton with hinged limbs.
  • Unbreakable plastic bones
  • Hinged mandible
  • Removable arms, legs, and skull
  • Natural movement in the joints wherever possible
  • Sectioned calvarium to allow for study of the cranial vault
  • Jaw with full set of teeth, three of which are removable for closer examination
  • Spinal column shows the spinal cord, nerve exits, vertebral arteries, and a herniated disk
Life-sized human skeleton model (170 cm / 5'7" tall) for anatomical studies, cast in durable plastic with lifelike details. Features natural movement in joints, with hinged mandible, removable arms, legs, and skull. Sectioned calvarium allows for study of cranial vault; jaw includes full set of teeth (three are removable for closer examination). Spinal column shows spinal cord, nerve exits, vertebral arteries, and a herniated disk.
The skeleton is mounted in an upright position on a heavy-duty base with smooth gliding casters. A dust cover and detailed key are included.
Specifications
  • Désignation:
    Human skeleton model, life-sized