Utility Tray, Polypropylene
470340-028
- Description:Utility tray, polypropylene
- Cat. no.:470340-028
A deep polypropylene tray designed for all labware needs.
- Autoclavable
- Deep 5" tray
- Tray dimensions: 15×14×5"
Steam autoclavable, polypropylene molded tray can be used for sterilizing and drying labware. Trays are also ideal for labware storage.