Draining Tray, Polypropylene
Draining tray, polypropylene
Catalog # 470340-140
Supplier: United Scientific Supplies
Supplier #: 78103
Jump to:
Product Details & Documents
Wide polypropylene draining tray.
- Autoclavable
- Polypropylene
- Wide net-like design
- Tray dimensions: 16×16×4"
This basket, with wide, net like structure on all sizes is useful for draining different laboratory apparatus after washing them. Made of polypropylene, this tray can be repeatedly autoclaved.
Specifications
- Désignation:Draining tray, polypropylene