Cryo Rack, Polycarbonate | Avantor
Cryo Rack, Polycarbonate
Cryo rack, polycarbonate
Catalog # 470339-404
Fournisseur #:  66201
Cryo Rack, Polycarbonate
Catalog # 470339-404
Cryo Rack, Polycarbonate
Cryo Rack, Polycarbonate
Cryo rack, polycarbonate
Price
33,80 $
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Durable and autoclavable cryo rack.
  • Anti-skid rubber feet
  • Alpha Numeric grid
  • Autoclavable
This durable and autoclavable Cryo rack can hold 50 vials. Unique design at the bottom of each well lets vials be unscrewed without removing them from the rack. The rack features an alpha numeric index, and anti-skid rubber feet.
Spécifications
  • Désignation:
    Cryo rack, polycarbonate