SmartDrop™ L Spectrophotometer
About this item
The SmartDrop™ series of nano spectrophotometers provides quick and easy quantification of small samples.
- Ideal for basic DNA/RNA/protein quantification and purity
- Affordable LED and silicon detector technology
- Fixed 0.5 mm path length, detect up to 2500 ng/µl of dsDNA
These spectrophotometers utilize a small sample volume which eliminates the need for dilutions, providing more accurate results. Each unit includes a full color, 7"
touchscreen with icon-driven menus and preprogrammed protocols gets users up and running quickly. No external computer is required.
SmartDrop™ L is the affordable choice for laboratories needing to determine nucleic acid and protein concentrations and purity of samples. The LED light source and silicon photocell sensor provide readings at 260 and 280 nm. Samples from 1.0 to 2.0 µl are loaded directly on the pedestal. Results are provided in about 5 seconds. The software automatically calculates and displays 260/280 ratios.
The resulting data can be printed, saved in the SmartDrop™'s onboard memory or exported via USB as an Excel file. All SmartDrop™ models include an integrated printer. For OD600 readings, each model includes a cuvette port and two optical glass cuvettes.
2 Options Available Below
Product Details & Documents
The SmartDrop™ series of nano spectrophotometers provides quick and easy quantification of small samples.
- Ideal for basic DNA/RNA/protein quantification and purity
- Affordable LED and silicon detector technology
- Fixed 0.5 mm path length, detect up to 2500 ng/µl of dsDNA
These spectrophotometers utilize a small sample volume which eliminates the need for dilutions, providing more accurate results. Each unit includes a full color, 7"
touchscreen with icon-driven menus and preprogrammed protocols gets users up and running quickly. No external computer is required.
SmartDrop™ L is the affordable choice for laboratories needing to determine nucleic acid and protein concentrations and purity of samples. The LED light source and silicon photocell sensor provide readings at 260 and 280 nm. Samples from 1.0 to 2.0 µl are loaded directly on the pedestal. Results are provided in about 5 seconds. The software automatically calculates and displays 260/280 ratios.
The resulting data can be printed, saved in the SmartDrop™'s onboard memory or exported via USB as an Excel file. All SmartDrop™ models include an integrated printer. For OD600 readings, each model includes a cuvette port and two optical glass cuvettes.
Product Family Options
115 V Unit
Product Information
- DésignationElectricalAvailabilityPrice
- Supplier Number: NS1000
230 V Unit
Product Information
- DésignationElectricalAvailabilityPrice
- Supplier Number: NS1000-E
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement printer roll