VWR® Cleanroom Binder
Cleanroom binder, Polyethylene, 11×17"
Catalog # 76547-914
Supplier: VWR International
VWR® Cleanroom Binder
Catalog # 76547-914
Supplier: VWR International
Jump to:
Product Details & Documents
Binder for use in cleanroom environments.
- 3-ring cleanroom binder
- Polyethylene construction
- Poly cover
This 2" cleanroom binder made from polyethylene features a metal 3-ring binder and a poly cover to reduce ESDs and low particulation.
Spécifications
- Désignation:Cleanroom binder
- Matière:Polyethylene
- Taille:11×17"
- Cat. No.:76547-914
- Environmentally Preferable:Sustainable Materials|Waste Reducing