PUREZERO* ULTRA VIOLET* Nitrile Exam Gloves, Halyard
Designed for use in laboratories, research environments, and clean areas with applications in pharmaceutical, medical device manufacturing, biotechnology, and food processing/handling.
- Features a SMARTPULL* Dispenser to control dispensing and lessen waste
- Powder-free and natural rubber latex-free
- Compliant for food handling applications
- AQL 1.0 (exceeds the ASTM standard of AQL 2.5 for critical defects or pinholes)
- Ambidextrous
- Beaded cuffs to prevent roll-down
Tested against 14 chemotherapy drugs, fentanyl, and 57 chemicals.
- Catalog No:
- 76554-030
- 76554-008
- 76554-038
- 76554-000
- 76554-026
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Finger thickness:
- 3.5 mil
- 3.5 mil
- 3.5 mil
- 3.5 mil
- 3.5 mil
- Palm thickness:
- 2.8 mil
- 2.8 mil
- 2.8 mil
- 2.8 mil
- 2.8 mil
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Frequently Bought Together