Estes Launch Lug Pack
Launch lugs pack (pack of 16)
Catalog # 470338-812
Supplier: Estes Industries
Fournisseur #: 2320
Jump to:
Product Details & Documents
Launch lug variety pack for Estes rocket body tubes.
- Contains four sizes of launch lugs (16 lugs total) in one convenient pack
- Can be used with ¹/₈", ³/₁₆" and ¹/₄" diameter launch rods
Contains four each. ¹/₈×1 ¹/₄", ³/₁₆× 2", ¹/₈× 2 ³/₈" and ¹/₄× 1" launch lugs.
Spécifications
- Désignation:Launch lugs pack (pack of 16)