À propos de cet article
Revolve™ sustainable wipes are made for the cleanroom industry and are composed of 100% upcycled polyester material with a sealed edge, processed on Texwipe’s fully automated manufacturing system. Revolve™ wipes contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
- Wipe and clean surfaces, equipment and parts
- Apply and remove lubricants, adhesives, residues and other solutions including disinfectants
- Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
Revolve™ processing provides low levels of ions, NVRs (non volatile residues), particles, and fibres for use in critical cleaning applications and environments. They are designed for high sorption capacity, which is ideal for spill control, cleaning, and solution application.
Revolve™ pre-wetted products are pre-wet with 0.2 µm filtered USP-grade 70% IPA/30% DIW. Pre-wetted wipes are packed in easy to use, recloseable slider bags, reducing solution evaporation that preserves the consistent wipes wetness level.
Spécifications
- Catalog No:
- 76472-844
- 76472-842
- 76472-840
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials