Revolve™ Sustainable Sterile Cleanroom Wipes, Texwipe®
Fournisseur: Texwipe
Certificats
À propos de cet article
Revolve™ sustainable wipes are made for the cleanroom industry and are composed of 100% upcycled polyester material with a sealed edge, processed on Texwipe’s fully automated manufacturing system. Revolve™ wipes contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
- Wipe and clean surfaces, equipment, and parts
- Apply and remove lubricants, adhesives, residues, and other solutions including disinfectants
- Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers
Revolve™ processing provides low levels of ions, NVRs (non-volatile residues), particles, and fibers for use in critical cleaning applications and environments. They are designed for high sorption capacity, which is ideal for spill control, cleaning, and solution application.
Certifications: Meet USP and USP wipe requirements. Sterile wipes are compatible for use in GMP grade A.
Spécifications
- Catalog No:
- 76472-838
- 76472-836
- 76472-834
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials