Masterflex® Ismatec® Microbore Tubing Straight Connectors, Avantor®
Fournisseur: VWR International
Certificats
À propos de cet article
Connect even the smallest tubing with a leak-free seal from vacuum to pressure.
- Multiple styles to fit your needs
- Stainless steel fitting sized to match the Ismatec® sizes - from 09 to 26
Delivery: Each kit includes 2 fittings.
- Environmentally Preferable:
Frequently Bought Together