KNF Remote-Control Diaphragm Liquid Dosing Pump Accessories
Replacement parts keep your dosing pump running smoothly.
- Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
Replacement parts keep your dosing pump running smoothly.
- Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
- Catalog No:
- MFLX78167-50
- MFLX78167-51
- MFLX78167-52
- MFLX78167-55
- MFLX78167-56
- MFLX78167-63
- MFLX78167-61
- MFLX78167-54
- MFLX78167-60
- MFLX78167-53
- Wetted materials:
- PVDF
- PEEK
- PVDF
- FEP
- FEP
- FFKM
- PTFE
- PEEK
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Product Family Options
- Désignation
- 1 year warranty
Specifications
Cat. No.MFLX78167-50Wetted materialsPVDFWarranty1 year1 year warranty - In-line Filter 35um PEEK for Dosing Pumps 78167-00 to -07-1 year warranty
Specifications
Cat. No.MFLX78167-51Wetted materialsPEEKWarranty1 year1 year warranty - 1 year warranty
Specifications
Cat. No.MFLX78167-52Wetted materialsPVDFWarranty1 year1 year warranty - Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 FittingEach$194.001 year warranty
Specifications
Cat. No.MFLX78167-55Wetted materialsFEPWarranty1 year1 year warranty - Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 FittingEach$342.001 year warranty
Specifications
Cat. No.MFLX78167-56Wetted materialsFEPWarranty1 year1 year warranty - 1 year warranty
Specifications
Cat. No.MFLX78167-63Wetted materialsFFKMWarranty1 year1 year warranty - Impulse Footswitch for Dosing Pumps 78167-00 to -10-1 year warrantyCE Compliant
Specifications
Cat. No.MFLX78167-61Wetted materialsWarranty1 year1 year warrantyCE Compliant - Connection Set UNF 1/4" for 1/8" OD tubing; 1m, 2 ferrules, 2 fittings-1 year warranty
Specifications
Cat. No.MFLX78167-54Wetted materialsWarranty1 year1 year warranty - Pressure Control Valve Set, PTFE, for Dosing Pumps 78167-00 to -07-1 year warranty
Specifications
Cat. No.MFLX78167-60Wetted materialsPTFEWarranty1 year1 year warranty - In-line Filter 35um PEEK for Dosing Pumps 78167-08 to -10-1 year warranty
Specifications
Cat. No.MFLX78167-53Wetted materialsPEEKWarranty1 year1 year warranty