PendoTECH® Single-Use Temperature Sensors
PendoTECH® Single-Use Temperature Sensors
Supplier:  PENDOTECH
Certificates
PendoTECH® Single-Use Temperature Sensors
5 Options Available Below
Product Details & Documents
Eliminates cross-contamination and extensive cleaning protocols.
  • Can be sterilized by gamma irradiation
  • High sensitivity to temperature change
  • Better than ±0.2 °C accuracy in specified range of 0 to 70 °C
  • Certificate of quality included
The PendoTECH single-use temperature sensors allow for highly accurate measuring of liquid temperature within a tube in a range of 0 to 70 °C. Their unique design allows part of the product that comes in contact with the fluid to be disposable. The sensors eliminate cross-contamination and extensive cleaning protocols. Although designed for single-use applications, the sensors are robust enough to be repeatedly cleaned and reused.
The sensors are available with either a hose barb or luer fitting for easy connection to process tubing. A disposable temperature dip probe is also available to measure temperature within a vessel or water bath. The hose barb models are constructed of polysulfone while the luer sensor is constructed of polycarbonate. The temperature sensing element in both sensors is a thermistor embedded in a stainless steel tip. No calibration is required because the temperature versus resistance for the thermistor element is well-defined within the specified accuracy range.
The hose barb fitting sensor imparts no obstruction on the fluid path that can cause a pressure drop and there is no dead-leg at the point where the temperature is measured. The luer fitting sensor is ideal for lab scale development applications and can connect to a variety of fittings that can securely adapt to tubing or other devices.
These sensors can be connected to a wide range of monitors and transmitters that can read the resistance measurement from the thermistor and convert it to a temperature reading. A reusable cable with a ¼” headphone plug (19406-15) can connect the sensor directly to a suitable monitor such as the Digi-Sense® handheld thermometer (91428-04) or a temperature sensor transmitter (19406-45). The temperature sensor transmitter with a linear 4-20 mA output signal allows a temperature reading to be displayed in another location such as process controller. To transmit temperature readings to a data logger, three benchtop transmitter options (19406-47, -57 and -58) are also available.
The sensors all have a three-year shelf life and all product contact materials pass the USP Class VI testing with the exception of the polycarbonate connector for luer sensor and the temperature sensor dip probe.
Specifications for Product Family
Specifications
  • Min. temperature:
  • 0 °C
  • 0 °C
  • 0 °C
  • 0 °C
  • 0 °C
  • 0 °C
  • Max. temperature:
  • 70 °C
  • 70 °C
  • 70 °C
  • 70 °C
  • 70 °C
  • 70 °C
  • Temperature accuracy:
  • Better than +/- 0.2 °C (typical better than 0.1 °C)
  • Better than +/- 0.2 °C (typical better than 0.1 °C)
  • Better than +/- 0.2 °C (typical better than 0.1 °C)
  • Better than +/- 0.2 °C (typical better than 0.1 °C)
  • Better than +/- 0.2 °C (typical better than 0.1 °C)
  • Better than +/- 0.2 °C (typical better than 0.1 °C)
  • Connector type:
  • 1/2" Hose Barb
  • 3/8" Hose Barb
  • 1/4" Hose Barb
  • 3/4" Hose Barb
  • 1/8" Hose Barb
  • Molex
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Specifications
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/2" Hose Barb
    Log in for availability
    Log in for availability
    Each
    Each - $185.00
    $185.00
    Each
       $185.00
    Supplier Number: TEMPS-N-050
    Specifications:
    2 year warranty
    CE certificate
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX19406-13
    Min. temperature
    0 °C
    Max. temperature
    70 °C
    Temperature accuracy
    Better than +/- 0.2 °C (typical better than 0.1 °C)
    Connector type
    1/2" Hose Barb
    Warranty
    2 years
    2 year warranty
    CE certificate
    CE Compliant
  • Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 3/8" Hose Barb
    Log in for availability
    Log in for availability
    Each
    Each - $175.00
    $175.00
    Each
       $175.00
    Supplier Number: TEMPS-N-038
    Specifications:
    2 year warranty
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX19406-16
    Min. temperature
    0 °C
    Max. temperature
    70 °C
    Temperature accuracy
    Better than +/- 0.2 °C (typical better than 0.1 °C)
    Connector type
    3/8" Hose Barb
    Warranty
    2 years
    2 year warranty
    CE certificate
  • Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/4" Hose Barb
    Log in for availability
    Log in for availability
    Each
    Each - $168.00
    $168.00
    Each
       $168.00
    Supplier Number: TEMPS-N-025
    Specifications:
    2 year warranty
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX19406-17
    Min. temperature
    0 °C
    Max. temperature
    70 °C
    Temperature accuracy
    Better than +/- 0.2 °C (typical better than 0.1 °C)
    Connector type
    1/4" Hose Barb
    Warranty
    2 years
    2 year warranty
    CE certificate
  • Single-Use Temperature Sensor, Non-Sterile, Polysulfone 3/4” Hose Barb
    Log in for availability
    Log in for availability
    Each
    Each - $174.00
    $174.00
    Each
       $174.00
    Supplier Number: TEMPS-N-075
    Specifications:
    2 year warranty
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX19406-42
    Min. temperature
    0 °C
    Max. temperature
    70 °C
    Temperature accuracy
    Better than +/- 0.2 °C (typical better than 0.1 °C)
    Connector type
    3/4" Hose Barb
    Warranty
    2 years
    2 year warranty
    CE certificate
  • Single-Use Temperature Sensor, Non-Sterile, Polysulfone 1/8” Hose Barb
    Log in for availability
    Log in for availability
    Each
    Each - $187.00
    $187.00
    Each
       $187.00
    Supplier Number: TEMPS-N-012
    Specifications:
    2 year warranty
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX19406-43
    Min. temperature
    0 °C
    Max. temperature
    70 °C
    Temperature accuracy
    Better than +/- 0.2 °C (typical better than 0.1 °C)
    Connector type
    1/8" Hose Barb
    Warranty
    2 years
    2 year warranty
    CE certificate
  • Single-Use Temperature Sensor Dip Probe Tip, Sterile, Molex Connector to Monitor Cable
    Discontinued
    Supplier Number: -
    Specifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    MFLX19406-44
    Min. temperature
    0 °C
    Max. temperature
    70 °C
    Temperature accuracy
    Better than +/- 0.2 °C (typical better than 0.1 °C)
    Connector type
    Molex
    Warranty
    2 years
    2 year warranty
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Temperature Sensor System Accessory with 4-20mA Output in ABS Plastic Box; 24VDC Wall Supply ***Must order cable separately***
    Temperature Sensor System Accessory with 4-20mA Output in ABS Plastic Box; 24VDC Wall Supply ***Must order cable separately***
    MFLX19406-47
    Log in for availability
    Each - $1,920.00
Product Information
  • Catalog #
    Availability
    Price
  • Reusable Temperature Sensor Cable with M8 Termination for Hose Barb Sensors; 12"
    Reusable Temperature Sensor Cable with M8 Termination for Hose Barb Sensors; 12"
    MFLX19406-48
    Log in for availability
    Each - $152.00
Product Information
  • Catalog #
    Availability
    Price
  • Reusable Temperature Sensor Cable with 1/4" Phone Jack for Luer Sensors; 7 ft (2 m)
    Reusable Temperature Sensor Cable with 1/4" Phone Jack for Luer Sensors; 7 ft (2 m)
    MFLX19406-14
    Log in for availability
    Each - $560.00
Product Information
  • Catalog #
    Availability
    Price
  • Reusable Temperature Sensor Cable with 1/4" Phone Jack for Hose Barb Fitting; 10 ft (3 m)
    Reusable Temperature Sensor Cable with 1/4" Phone Jack for Hose Barb Fitting; 10 ft (3 m)
    MFLX19406-15
    Log in for availability
    Each - $245.00
Product Information
  • Catalog #
    Availability
    Price
  • Adapter Tee, 1/4" Hose Barb x 1/4" Hose Barb x Male Luer Fitting, Polycarbonate
    Adapter Tee, 1/4" Hose Barb x 1/4" Hose Barb x Male Luer Fitting, Polycarbonate
    MFLX19406-34
    Log in for availability
    Each - $41.10
Product Information
  • Catalog #
    Availability
    Price
  • Adapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
    Adapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
    MFLX19406-35
    Log in for availability
    Each - $40.90
Product Information
  • Catalog #
    Availability
    Price
  • Adapter Tee, 3/8" Hose Barb x 3/8" Hose Barb x Male Luer Fitting, Polycarbonate
    Adapter Tee, 3/8" Hose Barb x 3/8" Hose Barb x Male Luer Fitting, Polycarbonate
    MFLX19406-36
    Log in for availability
    Each - $26.10
Product Information
  • Catalog #
    Availability
    Price
  • Temperature Sensor Transmitter with 4-20mA Output
    Temperature Sensor Transmitter with 4-20mA Output
    MFLX19406-45
    Log in for availability
    Each - $695.00
Product Information
  • Catalog #
    Availability
    Price
  • Temperature Sensor Transmitter DIN Rail Mounting Kit
    Temperature Sensor Transmitter DIN Rail Mounting Kit
    MFLX19406-46
    Log in for availability
    Each - $63.30