Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor® | Avantor
Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor®
Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor®
Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor®
Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor®
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing.
  • Constructed of durable Polypropylene
All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI. Available with 3/8" or 1/2" hose barbs and 3/4" mini or 1-1/2" standard sanitary clamp connections.
Documents
  • Sterilisation:
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • None
  • None
  • None
  • None
  • None
  • None
  • None
Product Family Options
  • Désignation
    Taille de l'ardillon/Tube Ø int. (port 2)
    Type de connection
    Flow pattern
    Sanitary clamp size (Port 1)
    Fitting Material
    Sanitary Gasket Size
  • Barbed to Sanitary Adapter, ⅜" ID to Mini ¾" ADCF-PP
    ⅜"
    Sanitary to Barb
    Straight
    ¾"
    PP
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-13
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    3 Months
    CE certificate
  • Barbed to Sanitary Adapter, ½" ID to Mini ¾" ADCF-PP
    ½"
    Sanitary to Barb
    Straight
    ¾"
    PP
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-14
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    3 Months
    CE certificate
  • Barbed to Sanitary Adapter, ⅜" ID to 1-½", ADCF-PP
    ⅜"
    Sanitary to Barb
    Straight
    1½"
    PP
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-15
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    3 Months
    CE certificate
  • Barbed to Sanitary Adapter, ½" ID to 1-½", ADCF-PP
    ½"
    Sanitary to Barb
    Straight
    1½"
    PP
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-16
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    3 Months
    CE certificate
  • Sanitary End Cap, Mini ¾", ADCF-PP
    Sanitary Cap
    ¾"
    PP
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-17
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    None
    CE certificate
  • Sanitary End Cap, ADCF Polypropylene; 1-½"
    Sanitary Cap
    Straight
    1½"
    PP
    1½"
    Each
    Each - $105.00
    $105.00
    Each
       $105.00
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-18
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    None
    CE certificate
  • Barbed to Sanitary Adapter, ¼" ID to Mini ¾" ADCF-PP
    ¼"
    Sanitary to Barb
    Straight
    ¾"
    PP
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-51
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    None
    CE certificate
  • Barbed to Sanitary Adapter, ¾" ID to Mini ¾" ADCF-PP
    ¾"
    Sanitary to Barb
    Straight
    ¾"
    PP
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-54
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    None
    CE certificate
  • Barbed to Sanitary Adapter, ¼" ID to 1-½", ADCF-PP
    ¼"
    Sanitary to Barb
    Straight
    1½"
    PP
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-55
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    None
    CE certificate
  • Barbed to Sanitary Adapter, ¾" ID to 1-½", ADCF-PP
    ¾"
    Sanitary to Barb
    Straight
    1½"
    PP
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-58
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    None
    CE certificate
  • Barbed to Sanitary Adapter, ADCF Polypropylene; 1" ID to 1-½"
    1"
    Sanitary to Barb
    Straight
    1½"
    PP
    1½"
    CE certificate
    Specifications
    Cat. No.
    MFLX31513-59
    Sterilisation
    Ethylene oxide (ETO) or gamma
    Warranty
    None
    CE certificate