Replacement PFA Compression Ferrule Nut and Assemblies
Replacement PFA Compression Ferrule Nut and Assemblies
Replacement PFA Compression Ferrule Nut and Assemblies
Certificates
Replacement PFA Compression Ferrule Nut and Assemblies
Replacement PFA Compression Ferrule Nut and Assemblies
5 Options Available Below
Product Details & Documents
Create a secure connection with compression fittings.
  • Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications for Product Family
Spécifications
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Warranty:
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Type de connection
    Flow pattern
    Fitting Material
    Availability
    Price
  • MFLX31320-46
    Ferrule plug, PFA, ¼"
    Compression Plug
    PFA
    Discontinued
    Spécifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-46
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • MFLX31321-85
    Compression Nut Assembly, PFA, ⅛"
    Compression Nut Assembly with Ferrule
    Straight
    PFA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $34.29
    $34.29
    Each
       $34.29
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX31321-85
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31321-86
    Compression Nut Assembly, PFA, ¼"
    Compression Nut Assembly with Ferrule
    Straight
    PFA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $34.29
    $34.29
    Each
       $34.29
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX31321-86
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31321-87
    Compression Nut Assembly, PFA, ⅜"
    Compression Nut Assembly with Ferrule
    Straight
    PFA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $41.24
    $41.24
    Each
       $41.24
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX31321-87
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31321-88
    Compression Nut Assembly, PFA, ½"
    Compression Nut Assembly with Ferrule
    Straight
    PFA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $44.56
    $44.56
    Each
       $44.56
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX31321-88
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31321-89
    Compression Nut Assembly, PFA, ¾"
    Compression Nut Assembly with Ferrule
    Straight
    PFA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $59.09
    $59.09
    Each
       $59.09
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX31321-89
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31320-45
    Ferrule plug, PFA, ⅛"
    Compression Plug
    PFA
    Discontinued
    Spécifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-45
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • MFLX31320-47
    Compression Ferrule Plug Fitting, PFA, ⅜"; 1/Each
    Compression Plug
    Straight
    PFA
    Discontinued
    Spécifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-47
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • MFLX31320-49
    Ferrule plug, PFA, ¾"
    Compression Plug
    PFA
    Discontinued
    Spécifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-49
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • MFLX31320-48
    Compression Ferrule Plug Fitting, PFA, ½"; 1/Each
    Compression Plug
    Straight
    PFA
    Discontinued
    Spécifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-48
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate