Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
35 Options Available Below
Product Details & Documents
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
Specifications for Product Family
Spécifications
  • Connection type:
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • No. of ports:
  • 2
  • 3
  • 4
  • 4
  • 1
  • 1
  • 1
  • 3
  • 1
  • 2
  • 3
  • 1
  • 1
  • 2
  • 4
  • 1
  • 2
  • 3
  • 3
  • 1
  • 4
  • 1
  • 1
  • 1
  • 1
  • 2
  • 2
  • 2
  • 2
  • 3
  • 3
  • 1
  • 1
  • 1
  • 2
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Sanitary clamp size (Port 1)
    Fitting Material
    Sanitary Gasket Size
    Availability
    Price
  • MFLX30543-13
    1 1/2" Tri-Clamp, 2 Port, EPDM, Each
    1½"
    EPDM
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $181.00
    $181.00
    Each
       $181.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30543-13
    Connection type
    Sanitary
    No. of ports
    2
    Warranty
    1 year
  • MFLX30544-14
    1 1/2" Tri-Clamp, 3 Port, Viton, Each
    1½"
    Viton®
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $185.00
    $185.00
    Each
       $185.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30544-14
    Connection type
    Sanitary
    No. of ports
    3
    Warranty
    1 year
  • MFLX30545-12
    1 1/2" Tri-Clamp, 4 Port, Silicone, Each
    1½"
    Platinum-cured silicone
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $255.00
    $255.00
    Each
       $255.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30545-12
    Connection type
    Sanitary
    No. of ports
    4
    Warranty
    1 year
  • MFLX30545-14
    1 1/2" Tri-Clamp, 4 Port, Viton, Each
    1½"
    Viton®
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $215.00
    $215.00
    Each
       $215.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30545-14
    Connection type
    Sanitary
    No. of ports
    4
    Warranty
    1 year
  • MFLX30542-09
    1" Tri-Clamp, 1 Port, EPDM, Each
    1"
    EPDM
    1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $154.00
    $154.00
    Each
       $154.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-09
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30542-10
    1" Tri-Clamp, 1 Port, Viton, Each
    1"
    Viton®
    1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $147.00
    $147.00
    Each
       $147.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-10
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30542-01
    1/2" Tri-Clamp Gasket, 1 Port, EPDM; 1/EA
    ¾"
    EPDM
    ½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $111.00
    $111.00
    Each
       $111.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-01
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30544-00
    1/2" Tri-Clamp, 3 Port, Silicone, Each
    ½"
    Platinum-cured silicone
    ½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $160.00
    $160.00
    Each
       $160.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30544-00
    Connection type
    Sanitary
    No. of ports
    3
    Warranty
    1 year
  • 1-1/2" Tri-Clamp, 1 Port, Silicone; 1/EA
    1½"
    Platinum-Cured Silicone
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $156.00
    $156.00
    Each
       $156.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-12
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30543-12
    1-1/2" Tri-Clamp, 2 Port, Silicone; 1/EA
    1½"
    Platinum-Cured Silicone
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.00
    $191.00
    Each
       $191.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30543-12
    Connection type
    Sanitary
    No. of ports
    2
    Warranty
    1 year
  • MFLX30544-12
    1-1/2" Tri-Clamp, 3 Port, Silicone; 1/EA
    1½"
    Platinum-Cured Silicone
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $210.00
    $210.00
    Each
       $210.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30544-12
    Connection type
    Sanitary
    No. of ports
    3
    Warranty
    1 year
  • MFLX30542-17
    2" Tri-Clamp, 1 Port, EPDM, Each
    2"
    EPDM
    2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $164.00
    $164.00
    Each
       $164.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-17
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30542-18
    2" Tri-Clamp, 1 Port, Viton, Each
    2"
    Viton®
    2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $171.00
    $171.00
    Each
       $171.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-18
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30543-18
    2" Tri-Clamp, 2 Port, Viton, Each
    2"
    Viton®
    2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $174.00
    $174.00
    Each
       $174.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30543-18
    Connection type
    Sanitary
    No. of ports
    2
    Warranty
    1 year
  • MFLX30545-16
    2" Tri-Clamp, 4 Port, Silicone, Each
    2"
    Platinum-cured silicone
    2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $254.00
    $254.00
    Each
       $254.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30545-16
    Connection type
    Sanitary
    No. of ports
    4
    Warranty
    1 year
  • MFLX30542-05
    3/4" Tri-Clamp Gasket, 1 Port, EPDM; 1/EA
    ¾"
    EPDM
    ¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $111.00
    $111.00
    Each
       $111.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-05
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30543-05
    3/4" Tri-Clamp, 2 Port, EPDM, Each
    ¾"
    EPDM
    ¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $123.00
    $123.00
    Each
       $123.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30543-05
    Connection type
    Sanitary
    No. of ports
    2
    Warranty
    1 year
  • MFLX30544-05
    3/4" Tri-Clamp, 3 Port, EPDM, Each
    ¾"
    EPDM
    ¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $147.00
    $147.00
    Each
       $147.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30544-05
    Connection type
    Sanitary
    No. of ports
    3
    Warranty
    1 year
  • MFLX30544-06
    3/4" Tri-Clamp, 3 Port, Viton, Each
    ¾"
    Viton®
    ¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $150.00
    $150.00
    Each
       $150.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30544-06
    Connection type
    Sanitary
    No. of ports
    3
    Warranty
    1 year
  • MFLX30542-13
    Sanitary Sampling Gasket, EPDM, 1 Port, 1-1/2" Tri-Clamp; 1/EA
    1½"
    EPDM
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $154.00
    $154.00
    Each
       $154.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-13
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30545-13
    Sanitary Sampling Gasket, EPDM, 4 Port, 1-1/2" Tri-Clamp; 1/EA
    1½"
    EPDM
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $241.00
    $241.00
    Each
       $241.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30545-13
    Connection type
    Sanitary
    No. of ports
    4
    Warranty
    1 year
  • MFLX30542-08
    Sanitary Sampling Gasket, Silicone, 1 Port, 1" Tri-Clamp; 1/EA
    1½"
    Platinum-Cured Silicone
    1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.00
    $175.00
    Each
       $175.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-08
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30542-00
    Sanitary Sampling Gasket, Silicone, 1 Port, 1/2" Tri-Clamp; 1/EA
    ¾"
    Platinum-Cured Silicone
    ½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $116.00
    $116.00
    Each
       $116.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-00
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30542-16
    Sanitary Sampling Gasket, Silicone, 1 Port, 2" Tri-Clamp; 1/EA
    2"
    Platinum-Cured Silicone
    2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $181.00
    $181.00
    Each
       $181.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-16
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30542-04
    Sanitary Sampling Gasket, Silicone, 1 Port, 3/4" Tri-Clamp; 1/EA
    ¾"
    Platinum-Cured Silicone
    ¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $121.00
    $121.00
    Each
       $121.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-04
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30543-08
    Sanitary Sampling Gasket, Silicone, 2 Port, 1" Tri-Clamp; 1/EA
    1½"
    Platinum-Cured Silicone
    1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.00
    $191.00
    Each
       $191.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30543-08
    Connection type
    Sanitary
    No. of ports
    2
    Warranty
    1 year
  • MFLX30543-00
    Sanitary Sampling Gasket, Silicone, 2 Port, 1/2" Tri-Clamp; 1/EA
    ¾"
    Platinum-Cured Silicone
    ½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $143.00
    $143.00
    Each
       $143.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30543-00
    Connection type
    Sanitary
    No. of ports
    2
    Warranty
    1 year
  • MFLX30543-16
    Sanitary Sampling Gasket, Silicone, 2 Port, 2" Tri-Clamp; 1/EA
    2"
    Platinum-Cured Silicone
    2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $209.00
    $209.00
    Each
       $209.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30543-16
    Connection type
    Sanitary
    No. of ports
    2
    Warranty
    1 year
  • MFLX30543-04
    Sanitary Sampling Gasket, Silicone, 2 Port, 3/4" Tri-Clamp; 1/EA
    ¾"
    Platinum-Cured Silicone
    ¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $149.00
    $149.00
    Each
       $149.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30543-04
    Connection type
    Sanitary
    No. of ports
    2
    Warranty
    1 year
  • MFLX30544-16
    Sanitary Sampling Gasket, Silicone, 3 Port, 2" Tri-Clamp; 1/EA
    2"
    Platinum-Cured Silicone
    2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $212.00
    $212.00
    Each
       $212.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30544-16
    Connection type
    Sanitary
    No. of ports
    3
    Warranty
    1 year
  • MFLX30544-04
    Sanitary Sampling Gasket, Silicone, 3 Port, 3/4" Tri-Clamp; 1/EA
    ¾"
    Platinum-Cured Silicone
    ¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $160.00
    $160.00
    Each
       $160.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30544-04
    Connection type
    Sanitary
    No. of ports
    3
    Warranty
    1 year
  • MFLX30542-02
    Sanitary Sampling Gasket, Viton, 1 Port, 1/2" Tri-Clamp; 1/EA
    ¾"
    Viton®
    ½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $113.00
    $113.00
    Each
       $113.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-02
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30542-14
    Sanitary Sampling Gasket, Viton, 1 Port, 1-1/2" Tri-Clamp; 1/EA
    1½"
    Viton®
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $147.00
    $147.00
    Each
       $147.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-14
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30542-06
    Sanitary Sampling Gasket, Viton, 1 Port, 3/4" Tri-Clamp; 1/EA
    ¾"
    Viton®
    ¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $113.00
    $113.00
    Each
       $113.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30542-06
    Connection type
    Sanitary
    No. of ports
    1
    Warranty
    1 year
  • MFLX30543-14
    Sanitary Sampling Gasket, Viton, 2 Port, 1-1/2" Tri-Clamp; 1/EA
    1½"
    Viton®
    1½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $168.00
    $168.00
    Each
       $168.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX30543-14
    Connection type
    Sanitary
    No. of ports
    2
    Warranty