Electrically and Pneumatically Actuated Elliptical Valves
Supplier:   EVSCO INC
Electrically and Pneumatically Actuated Elliptical Valves
Supplier:  EVSCO INC
Certificates
Electrically and Pneumatically Actuated Elliptical Valves
Electrically and Pneumatically Actuated Elliptical Valves
2 Options Available Below
Product Details & Documents
Completely disassemble in-line for easy cleaning.
  • FDA-compliant
  • Choose between Polypropylene (PP) or PVDF valve options
Use these actuated elliptical valves for liquid, gas, and vacuum applications. Valve systems are available in PP or PVDF bodies; all are FDA compliant and have O-rings made of Viton®. Rated for 150 psi at 73 °F (10 bar at 22 °C) (liquids).
Specifications for Product Family
Spécifications
  • Actuation motor:
  • 115V
  • Pneumatic
  • 115V
  • 115V
  • 115V
  • 115V
  • Pneumatic
  • Pneumatic
  • Pneumatic
  • Media compatibility:
  • Use for liquid, gas, and vacuumapplications
  • Use for liquid, gas, and vacuumapplications
  • Use for liquid, gas, and vacuumapplications
  • Use for liquid, gas, and vacuumapplications
  • Use for liquid, gas, and vacuumapplications
  • Use for liquid, gas, and vacuumapplications
  • Use for liquid, gas, and vacuumapplications
  • Use for liquid, gas, and vacuumapplications
  • Use for liquid, gas, and vacuumapplications
  • Orifice:
  • 19.05 mm (0.75")
  • 6.35 mm (0.25")
  • 6.35 mm (0.25")
  • 12.7 mm (0.5")
  • 3.175 mm (0.125")
  • 12.7 mm (0.5")
  • 19.05 mm (0.75")
  • 6.35 mm (0.25")
  • 19.05 mm (0.75")
  • Power:
  • 115 VAC
  • 115 VAC
  • 115 VAC
  • 115 VAC
  • 115 VAC
  • Warranty:
  • 1 Month
  • 1 Month
  • 1 Month
  • 1 Month
  • 1 Month
  • 1 Month
  • 1 Month
  • 1 Month
  • 3 Months
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Matériau du boîtier
    Type de connection
    Taille du port
    Inlet Port
    Outlet Port
    Availability
    Price
  • MFLX98153-16
    Three-Way Elliptical Valve, PP, 115 VAC, 3/4" NPT(F)
    PP
    NPT(F)
    0.75 NPT(F)
    ¾" NPT(F)
    ¾" NPT(F)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $719.00
    $719.00
    Each
       $719.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX98153-16
    Actuation motor
    115V
    Media compatibility
    Use for liquid, gas, and vacuumapplications
    Orifice
    19.05 mm (0.75")
    Power
    115 VAC
    Warranty
    1 Month
  • MFLX98154-13
    Three-Way Elliptical Valve, PP, Pneumatic, 1/4" NPT(F)
    PP
    NPT(F)
    0.25 NPT(F)
    ¼" NPT(F)
    ¼" NPT(F)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $664.00
    $664.00
    Each
       $664.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX98154-13
    Actuation motor
    Pneumatic
    Media compatibility
    Use for liquid, gas, and vacuumapplications
    Orifice
    6.35 mm (0.25")
    Power
    Warranty
    1 Month
  • MFLX98153-03
    Two-Way Elliptical Valve, PP, 115 VAC, 1/4" NPT(F)
    PP
    NPT(F)
    0.25 NPT(F)
    ¼" NPT(F)
    ¼" NPT(F)
    Discontinued
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX98153-03
    Actuation motor
    115V
    Media compatibility
    Use for liquid, gas, and vacuumapplications
    Orifice
    6.35 mm (0.25")
    Power
    115 VAC
    Warranty
    1 Month
  • MFLX98153-15
    Three-Way Elliptical Valve, PP, 115 VAC, 1/2" NPT(F)
    PP
    NPT(F)
    0.5 NPT(F)
    ½" NPT(F)
    ½" NPT(F)
    Discontinued
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX98153-15
    Actuation motor
    115V
    Media compatibility
    Use for liquid, gas, and vacuumapplications
    Orifice
    12.7 mm (0.5")
    Power
    115 VAC
    Warranty
    1 Month
  • MFLX98153-30
    Three-Way Elliptical Valve, PVDF, 115 VAC, 1/8" NPT(F)
    PVDF
    NPT(F)
    0.125 NPT(F)
    ⅛" NPT(F)
    ⅛" NPT(F)
    Discontinued
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX98153-30
    Actuation motor
    115V
    Media compatibility
    Use for liquid, gas, and vacuumapplications
    Orifice
    3.175 mm (0.125")
    Power
    115 VAC
    Warranty
    1 Month
  • MFLX98153-33
    Three-Way Elliptical Valve, PVDF, 115 VAC, 1/2" NPT(F)
    PVDF
    NPT(F)
    0.5 NPT(F)
    ½" NPT(F)
    ½" NPT(F)
    Discontinued
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX98153-33
    Actuation motor
    115V
    Media compatibility
    Use for liquid, gas, and vacuumapplications
    Orifice
    12.7 mm (0.5")
    Power
    115 VAC
    Warranty
    1 Month
  • MFLX98154-16
    Three-Way Elliptical Valve, PP, Pneumatic, 3/4" NPT(F)
    PP
    NPT(F)
    0.75 NPT(F)
    ¾" NPT(F)
    ¾" NPT(F)
    Discontinued
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX98154-16
    Actuation motor
    Pneumatic
    Media compatibility
    Use for liquid, gas, and vacuumapplications
    Orifice
    19.05 mm (0.75")
    Power
    Warranty
    1 Month
  • MFLX98154-31
    Three-Way Elliptical Valve, PVDF, Pneumatic, 1/4" NPT(F)
    PVDF
    NPT(F)
    0.25 NPT(F)
    ¼" NPT(F)
    ¼" NPT(F)
    Discontinued
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX98154-31
    Actuation motor
    Pneumatic
    Media compatibility
    Use for liquid, gas, and vacuumapplications
    Orifice
    6.35 mm (0.25")
    Power
    Warranty
    1 Month
  • MFLX98155-14
    Three-Way Elliptical Valve, PP, Pneumatic, 3/4" NPT(F)
    PP
    NPT(F)
    0.75 NPT(F)
    ¾" NPT(F)
    ¾" NPT(F)
    Discontinued
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX98155-14
    Actuation motor
    Pneumatic
    Media compatibility
    Use for liquid, gas, and vacuumapplications
    Orifice
    19.05 mm (0.75")
    Power
    Warranty
    3 Months