Flowline Switch-Tek Ultrasonic Level Switches
Fournisseur: FLOWLINE
Certificats
À propos de cet article
Compatible with Smart Trak and Switch-Pak installation fittings.
- CSA approved intrinsically safe for use in hazardous environments
- 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity
The LU10 series ultrasonic level switch is an excellent choice for a broad range of liquids, including those with light coating or scaling-type characteristics. Liquid examples include xylene, chromic acid and light cooking oil.
- Environmentally Preferable:
Spécifications
- Catalog No:
- MFLX23002-16
- MFLX23002-14
- Min. process temperature:
- -40 °F
- -40 °F
- Max. process temperature:
- 80 °C
- 80 °C
- Max. pressure:
- 150 psi
- 150 psi
- Relay output:
- SPST relay rated for 60 VA
- SPST relay rated for 60 VA
- Wetted materials:
- PFA
- Polypropylene
- Warranty:
- 1 year
- 1 year