Liquid-Solids Separator / Concentrator Units
À propos de cet article
Proven design utilizes centrifugal action to remove and concentrate solids.
- No screens, cartridges or filters to clean or replace
- Backflushing is not required - minimizing the loss of water and process fluids
- Unit has no moving parts - less maintenance down time required
In many processes, removal of solids can extend the usefulness of the process media and protect equipment from fouling and abrasive wear; regulatory benefits also exist in many applications. The basic design requires no electrical power and minimal piping, which makes cost payback on the installation quick.
The unit has a low and consistent pressure loss making process integration easy. Made from carbon steel, it is extremely durable and relatively inexpensive compared to other separation technologies.
4 Options Available Below
Product Details & Documents
Proven design utilizes centrifugal action to remove and concentrate solids.
- No screens, cartridges or filters to clean or replace
- Backflushing is not required - minimizing the loss of water and process fluids
- Unit has no moving parts - less maintenance down time required
In many processes, removal of solids can extend the usefulness of the process media and protect equipment from fouling and abrasive wear; regulatory benefits also exist in many applications. The basic design requires no electrical power and minimal piping, which makes cost payback on the installation quick.
The unit has a low and consistent pressure loss making process integration easy. Made from carbon steel, it is extremely durable and relatively inexpensive compared to other separation technologies.
Specifications for Product Family
Spécifications
- Catalog No:
- MFLX29995-02
- MFLX29995-06
- MFLX29995-12
- MFLX29995-18
- MFLX29995-04
- MFLX29995-08
- MFLX29995-10
- MFLX29995-14
- Filter type:
- Liquid-Solid Separator
- Liquid-Solid Separator
- Liquid-Solid Separator
- Liquid-Solid Separator
- Liquid-Solid Separator
- Liquid-Solid Separator
- Liquid-Solid Separator
- Liquid-Solid Separator
- Height:
- 50.8 cm (20")
- 76.2 cm (30")
- 85.41 cm (33.625")
- 152.4 cm (60")
- 50.8 cm (20")
- 76.2 cm (30")
- 76.2 cm (30")
- 93.98 cm (37")
- Inlet connection:
- 1/2" NPT(M)
- 1" NPT(M)
- 2" NPT(M)
- 3" NPT(M)
- 3/4" NPT(M)
- 1-1/4" NPT(M)
- 1-1/2" NPT(M)
- 2-1/2" NPT(M)
- Max. particulate size:
- 0.25"
- 0.25"
- 0.375"
- 0.375"
- 0.25"
- 0.375"
- 0.375"
- 0.375"
- Max. pressure:
- 10 bar (150 psi)
- 10 bar (150 psi)
- 10 bar (150 psi)
- 10 bar (150 psi)
- 10 bar (150 psi)
- 10 bar (150 psi)
- 10 bar (150 psi)
- 10 bar (150 psi)
- Outlet connection:
- 1/2" NPT(M)
- 1" NPT(M)
- 2" NPT(M)
- 3" NPT(M)
- 3/4" NPT(M)
- 1-1/4" NPT(M)
- 1-1/2" NPT(M)
- 2-1/2" NPT(M)
- Pressure loss:
- 12 psi
- 12 psi
- 12 psi
- 12 psi
- 12 psi
- 12 psi
- 12 psi
- 12 psi
- Purging port:
- 3/4" NPT(M)
- 3/4" NPT(M)
- 3/4" NPT(M)
- 3/4" NPT(M)
- 3/4" NPT(M)
- 3/4" NPT(M)
- 3/4" NPT(M)
- 3/4" NPT(M)
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Spécifications
Product Family Options
Informations sur le produit
- DésignationMatièreØCollection Chamber CapacityFlow RangeAvailabilityPrice
- Liquid-solids separator/concentrator,1/2" inlet/outlet, 4 to 10 gal./minCarbon steel10.16 cm0.3 gal.4 - 10 gal./minLog in pour la disponibilitéEach$1,500.00Spécifications:
More
Specifications
Cat. No.MFLX29995-02Filter typeLiquid-Solid SeparatorHeight50.8 cm (20")Inlet connection1/2" NPT(M)Max. particulate size0.25"Max. pressure10 bar (150 psi)Outlet connection1/2" NPT(M)Pressure loss12 psiPurging port3/4" NPT(M)Warranty1 year - Liquid-solids separator/concentrator,1" inlet/outlet, 19 to 32 gal./minCarbon steel11.11 cm0.3 gal.19 - 32 gal./minLog in pour la disponibilitéEach$1,800.00Spécifications:
More
Specifications
Cat. No.MFLX29995-06Filter typeLiquid-Solid SeparatorHeight76.2 cm (30")Inlet connection1" NPT(M)Max. particulate size0.25"Max. pressure10 bar (150 psi)Outlet connection1" NPT(M)Pressure loss12 psiPurging port3/4" NPT(M)Warranty1 year - Liquid/Solids In-Line Separator, 65 to 108 gal./minCarbon steel13.97 cm0.8 gal.65 - 108 gal./minLog in pour la disponibilitéEach$2,780.00Spécifications:
More
Specifications
Cat. No.MFLX29995-12Filter typeLiquid-Solid SeparatorHeight85.41 cm (33.625")Inlet connection2" NPT(M)Max. particulate size0.375"Max. pressure10 bar (150 psi)Outlet connection2" NPT(M)Pressure loss12 psiPurging port3/4" NPT(M)Warranty1 year - Liquid/Solids In-Line Separator, 148 to 290 gal./min, with Three 1" ID Pipe LegsCarbon steel20.96 cm2.2 gal.148 - 290 gal./minLog in pour la disponibilitéEach$6,700.00Spécifications:
More
Specifications
Cat. No.MFLX29995-18Filter typeLiquid-Solid SeparatorHeight152.4 cm (60")Inlet connection3" NPT(M)Max. particulate size0.375"Max. pressure10 bar (150 psi)Outlet connection3" NPT(M)Pressure loss12 psiPurging port3/4" NPT(M)Warranty1 year - Liquid/Solids In-Line Separator, 10 to 20 gal./minCarbon steel10.16 cm0.3 gal.10 - 20 gal./minDiscontinued-Spécifications:
More
Specifications
Cat. No.MFLX29995-04Filter typeLiquid-Solid SeparatorHeight50.8 cm (20")Inlet connection3/4" NPT(M)Max. particulate size0.25"Max. pressure10 bar (150 psi)Outlet connection3/4" NPT(M)Pressure loss12 psiPurging port3/4" NPT(M)Warranty1 year - Liquid/Solids In-Line Separator, 28 to 48 gal./minCarbon steel11.11 cm0.3 gal.28 - 48 gal./minDiscontinued-Spécifications:
More
Specifications
Cat. No.MFLX29995-08Filter typeLiquid-Solid SeparatorHeight76.2 cm (30")Inlet connection1-1/4" NPT(M)Max. particulate size0.375"Max. pressure10 bar (150 psi)Outlet connection1-1/4" NPT(M)Pressure loss12 psiPurging port3/4" NPT(M)Warranty1 year - Liquid/Solids In-Line Separator, 45 to 70 gal./minCarbon steel11.11 cm0.3 gal.45 - 70 gal./minDiscontinued-Spécifications:
More
Specifications
Cat. No.MFLX29995-10Filter typeLiquid-Solid SeparatorHeight76.2 cm (30")Inlet connection1-1/2" NPT(M)Max. particulate size0.375"Max. pressure10 bar (150 psi)Outlet connection1-1/2" NPT(M)Pressure loss12 psiPurging port3/4" NPT(M)Warranty1 year - Liquid/Solids In-Line Separator, 95 to 155 gal./minCarbon steel15.88 cm1.25 gal.95 - 155 gal./minDiscontinued-Spécifications:
More
Specifications
Cat. No.MFLX29995-14Filter typeLiquid-Solid SeparatorHeight93.98 cm (37")Inlet connection2-1/2" NPT(M)Max. particulate size0.375"Max. pressure10 bar (150 psi)Outlet connection2-1/2" NPT(M)Pressure loss12 psiPurging port3/4" NPT(M)Warranty1 year
Accessories
Informations sur le produit
- Catalog #Compatible withAvailabilityPrice
- Mounting Bracket Kit for Liquid/Solids In-Line Separators, 1/2" to 3/4" Inlet/OutletMounting Bracket Kit for Liquid/Solids In-Line Separators, 1/2" to 3/4" Inlet/OutletMFLX29995-5029995-00, 29995-02, 29995-04Log in pour la disponibilitéEach - $109.00
Informations sur le produit
- Catalog #Compatible withAvailabilityPrice
- Mounting Bracket Kit for Liquid/Solids In-Line Separators, 1Mounting Bracket Kit for Liquid/Solids In-Line Separators, 1MFLX29995-5229995-06, 29995-08, 29995-10Log in pour la disponibilitéEach - $146.00
Informations sur le produit
- Catalog #Compatible withAvailabilityPrice
- Manual Purge Kit for Liquid/Solids In-Line SeparatorsManual Purge Kit for Liquid/Solids In-Line SeparatorsMFLX29995-6029995 series liquid-solid separators/concentratorsLog in pour la disponibilitéEach - $182.00
Informations sur le produit
- Catalog #Compatible withAvailabilityPrice
- Mounting Bracket Kit for Liquid/Solids In-Line Separators, 2Mounting Bracket Kit for Liquid/Solids In-Line Separators, 2MFLX29995-5429995-12, 29995-14Log in pour la disponibilitéEach - $226.00