Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Gold nanoparticle demonstration
- Cat. no.:470336-160
- Includes:Instructional manual and SDS booklet
- No. of Uses:5
Specifications
About this item
Perform a reduction reaction to understand and demonstrate the properties of gold nanoparticles.
- Understand and demonstrate properties of nanomaterials
- Aligned to NGSS practices
Explore the interesting properties of nanomaterials by reducing a solution of gold (III) chloride to form colloidal gold nanoparticles. As the reaction occurs, the mixture undergoes multiple color changes before becoming the bright red color characteristic of small gold nanoparticles. There are enough materials to complete the demonstration five times.