Logic® Vue Class II Enclosures, for Protection and Containment for Large Instruments, Labconco
Fournisseur: Labconco
Certificats
À propos de cet article
The Logic® Vue is a specialized Class II enclosure providing personnel and product protection for large automated instruments such as liquid handlers. The Logic® Vue provides laminar ISO 5 air.
- Large capacity enclosure
- Accommodates diverse range of instruments such as liquid handlers and cell sorters
- Provides accessibility from front and back
- Visibility from all sides
- Powered by MyLogic™ OS to interface with a variety of devices for additional researcher safety
Certifications: Meets NSF/ANSI Standard 49.
Informations sur les accessoires: 76545-604 Canopy Connection for thimble exhausting Logic Vue Class II Enclosures. Canopy maintains a negative pressure connection to house exhaust.
76545-606 The computer arm attaches directly to left side of Logic Vue and supports monitor, keyboard and mouse.
76545-608 Connects two Logic Vue Class II Enclosures together for an expanded work area. Connection provides a passthrough that is 38.9H×28D×10"W.
76545-606 The computer arm attaches directly to left side of Logic Vue and supports monitor, keyboard and mouse.
76545-608 Connects two Logic Vue Class II Enclosures together for an expanded work area. Connection provides a passthrough that is 38.9H×28D×10"W.
Packaging: The international packaging indicated for selected units includes reduced height for high cube shipping containers.
Caution: Installation required.
Spécifications
- Catalog No:
- 76545-600
- 76545-598
- 76545-592
- 76545-596
- 76545-594
- 76545-602
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Environmental certification:
- 78% recycled content
- 78% recycled content
- 78% recycled content
- 78% recycled content
- 78% recycled content
- 78% recycled content