Phosphate Activator Solution, CHEMetrics
Phosphate activator solution, 6 × 10 ml, Bottles
  CXA8500
 :   CHEMetrics
 :  A-8500
Phosphate Activator Solution, CHEMetrics
  CXA8500
 :  
Phosphate Activator Solution, CHEMetrics
Phosphate Activator Solution, CHEMetrics
Phosphate activator solution, 6 × 10 ml, Bottles
 :  
$40.86
Pack of 6
The Phosphate activator solution accessory pack (A-8500) supplies enough solution to perform at least 200 tests.
 : Shelf Life: 20 months
  • Type de conditionnement:
    Bottles
  • Désignation:
    Phosphate activator solution
  • Taille:
    6 × 10 ml