Iron Activator Solution, CHEMetrics | Avantor
Iron Activator Solution, CHEMetrics
Iron Activator Solution, 6× 10 ml, Bottles
Catalog # CXA-6000
Supplier:   CHEMetrics
Fournisseur #:  A-6000
Iron Activator Solution, CHEMetrics
Catalog # CXA-6000
Supplier:  CHEMetrics
CAS Number:  
Iron Activator Solution, CHEMetrics
Iron Activator Solution, CHEMetrics
Iron Activator Solution, 6× 10 ml, Bottles
CAS Number:  
Price
$64.88
Log in pour la disponibilité
Pack of 6
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The A-6000 accessory pack supplies enough Iron solution to perform at least 200 CHEMets tests, 200 Vacu-vials tests, or 42 VACUettes tests.
The Activator Solution is required for total iron analysis only.
Spécifications
Spécifications
  • Type de conditionnement:
    Bottles
  • Désignation:
    Iron Activator Solution
  • Taille:
    6× 10 ml