Sentinel XL® CBRN Conversion Kit - HP or HD PAPR System to CBRN PAPR System, ILC Dover
76533-632
- Description:Sentinel XL® CBRN conversion kit
- Cat. no.:76533-632
Convert an HP PAPR System into a CBRN PAPR System in a few easy steps.
The blower stays the same, only the hood and filters must be changed. Kit includes Butyl Hood Assembly and 3 CBRN cartridge filters.