Sentinel XT® Clear PAPR System with S-3112 Hood and Sentinel XT® Blower, ILC Dover
Catalog # 76533-598
Supplier: ILC DOVER LP
Specifications
- Description:Sentinel XT® clear PAPR system
- Size:Universal
- Cat. no.:76533-598
Specifications
About this item
The Sentinel XT® PAPR system offers up to 8cfm, 33% greater airflow within the hood and suit. The hood attaches to protective clothing with clips, for a secure fit.
- 320 degree unobstructed field of view
- LiPo rechargeable battery lasts up to 12 hours
- Ergonomically designed for comfort during long shifts and complex operations
- One size fits all
- Latex-free
The Sentinel XT® Clear PAPR System includes the S-3112 clear full hood, Sentinel XT® blower, 33" breathing tube, plug, LiPo rechargeable battery and charging cradle, Quick Loc belt, and 2 HEPA cartridge filters.